Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heptane (mixture of isomers) 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger

H2198-2.5L
BJH2198-2.5LEA 306.34 USD InStock
BJH2198-2.5L
Heptane (mixture of isomers) 99%, Reagent Grade
Heptane (mixture of isomers)
  • ≥95% for general laboratory use.

Beilstein Number: 1730763


Formula: H₃C(CH₂)₅CH₃
MW: 100.2 g/mol
Boiling Pt: 6.7 %(V)
Melting Pt: -90.5 °C
Density: 0.69 g/cm³ (15 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/73/704373.jpg
Order Now
Specification Test Results
Assay (GC) Min. 99 %
Refractive index (n 20/D) 1.386 - 1.388
IR spectrum complying
Appearance complying
Vapor Pressure 40 mmHg ( 20 °C)
Vapor Pressure 83 mmHg ( 37.7 °C)
Vapor Density 3.5 (vs air)
Explosion Limit 7 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty