Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol absolute acetone-free

Supplier: Honeywell Research Chemicals
Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

M1775-1GA
BJM1775-1GAEA 178.01 USD InStock
BJM1775-1GA
Methanol absolute acetone-free
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64-65 °C (1.013 hPa)
Melting Pt: -98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Vapor Pressure 97.68 mmHg ( 20 °C)
Appearance complying
Autoignition Temperature 725 °F
Refractive Index n20/D 1.329 (lit.)
Vapor Pressure 410 mmHg ( 50 °C)
Explosion Limit 36 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty