Would you like to log in again?
Pierce™ Protein Conjugate, Horseradish Peroxidase, Biotinylated, Thermo Scientific
Supplier: Thermo Scientific
Pierce
29139 29139
PI29139EA 266.22 USD InStock
PI29139 CAPI29139
Pierce™ Protein Conjugate, Horseradish Peroxidase, Biotinylated, Thermo Scientific
Assays Protein Assays
Biotinylated horse radish peroxidase is used as control or signal amplification in IHC to amplify the signal of biotinylated primary antibodies using the avidin-biotin complex staining method.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty