Would you like to log in again?
Inoculating Loops and Needles, Sterile, SARSTEDT
Supplier: Sarstedt
86.1562.050 86.1567.050 86.1562.010 86.1569.005 86.1569.001 86.1568.010 86.1567.010 86.1568.050
103574-402PK 96.45 USD InStock
103574-402 103575-126 102471-702 101093-921 101093-923 101093-919 101093-674 103574-404
Inoculating Loops and Needles, Sterile, SARSTEDT
Spreaders Inoculating Loops and Needles
These gamma-irradiated inoculation loops and needles are made of polystyrene.
  • Color coded for easy identification
  • Provides enhanced safety and minimizes cross contamination
Inoculation needles are suitable for the transfer of individual colonies while inoculation loops facilitate smears or inoculation in liquid media.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty