Would you like to log in again?
DBI-SALA Advanced 5-Piece Hoist System, ORS Nasco
Supplier: ORS Nasco
DBI-SALA®
098-8518000
300058-335EA 9115.4 USD InStock
300058-335
DBI-SALA Advanced 5-Piece Hoist System, ORS Nasco
Fall Protection Fall Arresters
This lightweight aluminum davit is very portable, breaking down into five easy to manage pieces
  • Complete with two pulleys for winches/SRLs anchor point and adjustable feet
  • Provides 12–29" offset and swings 180° side to side
Complete assembly sets up in minutes. This line is also available with many permanent base options as well as other offsets and height adjustable components.
Certifications: Meets or Exceeds all Industry Regulations Including OSHA and ANSI.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty