Would you like to log in again?
Fit-Check Banana Oil Ampules, Allegro®
Supplier: Allegro
037-0201
300056-772PK 24.82 USD InStock
300056-772
Fit-Check Banana Oil Ampules, Allegro®
Ampules
Provides a fast, safe, and easy method to perform qualitative respirator fit check.
When activated, the ampules give off a non-irritating odor of banana oil (Iso Amyl Acetate). If banana odor is detected by the wearer, the respirator must be adjusted or replaced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty