Would you like to log in again?
Graphite Electrode, Counter, Thermo Scientific
Supplier: Thermo Scientific
014754.KN 014751.KN 014746.KN
AA14754-KNEA 352.29 USD InStock
AA14754-KN AA14746-KN AA14751-KN
Graphite Electrode, Counter, Thermo Scientific
Electrodes Other Electrodes
Graphite electrodes are used as homogeneous catalysts
  • AGKSP grade
  • Ultra F purity
  • Light sensitive and hygroscopic
  • Incompatible with oxidizing agents
  • Applications include piston rings, thrust bearings, journal bearings and vanes
These are widely used in electric arc furnaces (EAF) and ladle furnaces (LF) for the melting of steel, non-ferrous metals and some non-metallic minerals. Carbon based seals are used in the shafts and fuel pumps of many aircraft jet engines.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty