Would you like to log in again?
Weiler® Stem-Mounted Narrow Conflex Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-17616 804-17610 804-17641 804-17615 804-17617 804-17609 804-17614 804-17618 804-17611
300010-188EA 25.4 USD InStock
300010-188 300010-189 300047-134 300010-182 300010-183 300010-184 300010-185 300010-186 300010-187
Weiler® Stem-Mounted Narrow Conflex Brush, ORS Nasco
Brushes
Designed for light duty and general-purpose applications. Use to remove rust, paint, oxides, and scale.
  • Narrow face
  • For light-duty brushing action
Can be used with power drills, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty