Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-L(+)-glutamate monohydrate ≥99%, white powder cell culture reagent

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms: L(+)-α-Glutamic acid sodium salt monohydrate(S)-(+)-2-Aminopentanedioic acid sodium salt monohydrateL(+)-Glutamic acid sodium salt monohydrateSodium L-Glutamate Monohydrate(S)-(+)-Sodium glutamate monohydrateL(+)-2-Aminopentanedioic acid sodium salt monohydrate(S)-(+)-Glutamic acid sodium salt monohydrate(S)-(+)-α-Glutamic acid sodium salt monohydrateL(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrateL(+)-Glutamic acid monosodium salt monohydrate(S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate

0219467780 0219467791 0219467790
76177-776EA 87.78 USD InStock
76177-776 76177-778 76177-726
Sodium-L(+)-glutamate monohydrate ≥99%, white powder cell culture reagent
Sodium-L(+)-glutamate monohydrate
L-Glutamic Acid is a non-essential amino acid for human development; referred to as an excitatory amino acid (EAA) due to its role in neurotransmission. Acts as a neurotransmitter at fast synapses; agonist at kainate, NMDA and quisqualate receptors. L-Glutamic acid is one of the two amino acids that contains a carboxylic acid group in its side chains.

L-Glutamic acid is used in cell culture as a component of MEM non-essential amino acids solution.
Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.


Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Melting Pt: 232 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
Optical Rotation (c=1, 1 N HCl) +20° to +36°
Loss on Drying <0.5%
pH (5%, aq soln) 6.7-7.2
Solubility (10% aq soln) Clear and Colorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty