Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Proto® Metric Impact Socket Bits, ORS Nasco
Supplier: ORS Nasco
577-7441-14M 577-7441-8M 577-7290-6M 577-7290-10M 577-7441-17M 577-7290-8M 577-7441-10M 577-7441-12M 577-7290-4M 577-7290-5M 577-7441-6M
300027-696EA 22.86 USD InStock
300027-696 300028-012 300027-698 300027-688 300028-010 300003-152 300003-150 300028-018 300027-692 300027-694 300028-014
Proto® Metric Impact Socket Bits, ORS Nasco
Tool Sets
  • Heat-treated
  • Precision machined tip for proper screw engagement
  • Measurements provided in metric
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty