Would you like to log in again?
Proto® Drive Impact Universal Joint, ORS Nasco
Supplier: ORS Nasco
577-07570A 577-10670A
300092-776EA 245.37 USD InStock
300092-776 300092-774
Proto® Drive Impact Universal Joint, ORS Nasco
Tool Sets
Features two through holes and a groove to accept a Proto® Retaining Ring or pin and O-ring to help secure the socket to the drive tool anvil
  • Tight tolerances for snug fit on fastener
  • Ball and spring design which allows for a 30° working angle
  • Designed to withstand the high torque and force associated with impact tools in repetitive applications
  • Smooth 360° rotation
Position at appropriate angle to turn fasteners where obstructions prevent a straight application. Used wherever powered drive tools are used to install or remove fasteners.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty