Would you like to log in again?
Human Cathepsin B (from Liver)
Supplier: EMD Millipore
219364-5U
80050-892EA 318.33 USD InStock
80050-892
Human Cathepsin B (from Liver)
Proteins and Peptides
Native cathepsin B from human liver, purified by affinity chromatography.Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
  • Buffer: 50 mM sodium acetate, 1 mM EDTA, pH 5.0
The most investigated enzyme of all lysosomal cysteine proteases. Upregulation and secretion of cathepsin B have been shown to occur in many types of tumors and to correlate positively with their invasive and metastatic capabilities. Cathepsin B is shown to facilitate tumor invasion by dissolving extracellular barriers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty