Would you like to log in again?
Omnifit® EZ Chromatography Columns with One Fixed and One Adjustable Endpieces, Diba Industries
Supplier: Diba Industries
Omnifit®
11943-12 11943-66 11943-48 11943-84 11943-33 11943-51 11942-82 11942-97 11943-69 11943-39 11943-57 11943-78 11943-03 11943-06 11943-72 11943-00 11942-94 11943-60 11943-45 11943-15 11942-73 11943-09 11943-63 11943-18 11943-75 11943-54 11943-42 11943-81 11942-91 11943-21 11943-36 11942-88 11943-24 11943-27 11942-85 11943-30 11942-76 11942-79
76175-738EA 789.35 USD InStock
76175-738 76175-858 76175-978 76175-834 76176-060 76175-756 76175-810 76175-732 76175-798 76175-852 76175-774 76175-750 76175-792 76176-104 76176-048 76176-248 76176-066 76176-242 76176-102 76176-084 76175-828 76175-804 76175-846 76176-072 76175-768 76175-822 76175-744 76176-090 76175-786 76175-840 76175-762 76175-780 76176-236 76176-078 76176-110 76176-096 76176-054 76175-816
Omnifit® EZ Chromatography Columns with One Fixed and One Adjustable Endpieces, Diba Industries
Chromatography Columns
Ergonomically designed endpieces provide simple and quick height adjustment.
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Delivery information: Delivered with two (0.5 and 2 m length) ¹/₁₆" Ext.Ø PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 µm PE, 10 µm PTFE, and 30 µm PTFE frits), and a Click-N-Seal® 10-32 PEEK conical port fitting.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty