Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SGSH Rabbit Polyclonal Antibody

Supplier: EMD Millipore
345886-5GM
80056-536EA 568.26 USD InStock
80056-536
Anti-SGSH Rabbit Polyclonal Antibody
Antibodies
This antibody was developed against Recombinant Protein corresponding to amino acids: CEKFGNGESGMGRIPDWTPQAYDPLDVLVPYFVPNTPAARADLAAQYTTVGRMDQGVGLVLQELRDAGVLNDTLVIFTSDNGIPFPSGRTNLYWPGTAEPL.

  • Western Blot 1:100-1:500
  • Immunohistochemistry
  • Immunohistochemistry-Paraffin 1:200-1:500
  • Preservative: 0.02% Sodium Azide

Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.

Type: Primary
Antigen: SGSH
Clonality: Polyclonal
Clone:
Conjugation: Unconjucated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty