Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium tetrathionate

Supplier: Pfaltz & Bauer

P23970-25G
101180-784EA 609.55 USD InStock
101180-784
Potassium tetrathionate
Potassium tetrathionate


Formula: K₂O₆S₄
MW: 302.46 g/mol
Density: 2,96 g/cm³
MDL Number: MFCD00079233
CAS Number: 13932-13-3
EINECS: 237-702-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty