Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD23 Ab-1 Mouse Monoclonal Antibody [clone: 1B12]

Supplier: Richard-Allan Scientific
MS-729-S
100155-294EA 1095.72 USD InStock
100155-294
Anti-CD23 Ab-1 Mouse Monoclonal Antibody [clone: 1B12]
Antibodies
CD23 is identical to low affinity IgE receptor found on B cells. CD23 is expressed on a subpopulation of peripheral blood cells, B lymphocytes and on EBV transformed B lymphoblastoid cell lines. CD23 is also detected in neoplastic cells from cases of B cell chronic lymphocytic leukemia and some cases on centroblastic/centrocytic lymphoma.

Type: Primary
Antigen: CD23 Ab-1
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjucated
Epitope:
Host: Murine
Isotype: IgG2a kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty