Would you like to log in again?
Weiler® Copper Center™ Tampico Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-17453 804-20180
300048-456CS 191.8 USD InStock
300048-456 300045-920
Weiler® Copper Center™ Tampico Wheel, ORS Nasco
Brushes
Tampico fill has excellent liquid retention and is highly resistant to heat, solvents, and chemicals.
  • Brushes are well suited for lighter applications and greater flexibility
  • Can be used for delicate scratch-free brushing
Designed for wet or dry cleaning and scrubbing, removing surface imperfections, polishing, and surface finishing. These brushes can be used with die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty