Would you like to log in again?
Weiler® Dualife® Twist Knot Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-08156
300042-780EA 174.37 USD InStock
300042-780
Weiler® Dualife® Twist Knot Wire Wheel, ORS Nasco
Brushes
These brushes can be used with right-angle grinders, straight grinders, and bench/pedastal grinders.
  • Rugged knot-type construction
  • Aggressive, high-impact action for severe applications
Designed for deburring, cleaning prior to welding, roughening, weld scale and corrosion removal, and rust and paint removal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty