Would you like to log in again?
Weiler® Nylox® Basic Section Wheel Brush,ORS Nasco
Supplier: ORS Nasco
Weiler®
804-20600
300045-788CS 159.37 USD InStock
300045-788
Weiler® Nylox® Basic Section Wheel Brush,ORS Nasco
Brushes
Designed for light duty deburring and finishing in non-production applications.
  • Low volume manual deburring
  • General use in a tool room or shop
  • Crimped filament
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty