Would you like to log in again?
Weiler® Plater's Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-22241
300044-616EA 242.07 USD InStock
300044-616
Weiler® Plater's Wire Wheel, ORS Nasco
Brushes
Ideal for antique or fine satin finishes on precious and non-ferrous metals.
Can be used with bench/pedastal grinders.
Wheels are also available in carbon steel, stainless steel, and nickel silver wire.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty