Would you like to log in again?
Weiler® Polyflex® Stem-Mounted Conflex Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-35310 804-35315
300046-696EA 555.1 USD InStock
300046-696 300046-698
Weiler® Polyflex® Stem-Mounted Conflex Brush, ORS Nasco
Brushes
Elastomer gradually wears exposing consistent short trim for maximum aggression and long life.
Use to remove rust, paint, oxides, and scale. Can be used with power drills, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty