Would you like to log in again?
Weiler® Crimped Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-35216
300043-656EA 210.74 USD InStock
300043-656
Weiler® Crimped Wire Wheel, ORS Nasco
Brushes
Heat stabilized elastomer gradually wears exposing consistent short trim for maximum aggression and long life.
  • Controlled brushing action
  • Can be used with straight grinders and right-angle grinders
Designed for weld cleaning, and the removal of slag, spatter, and oxidation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty