Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Crk p38 Polyclonal Antibody

Supplier: Boster
PB9101
10206-672EA 808.99 USD InStock
10206-672
Anti-Crk p38 Polyclonal Antibody
Antibodies
Polyclonal antibody for CRK detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CRK information: Molecular Weight: 33831 MW; Subcellular Localization: Cytoplasm . Cell membrane . Translocated to the plasma membrane upon cell adhesion.

Type: Primary
Antigen: Crk p38
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty