Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cathepsin D Polyclonal Antibody

Supplier: Boster

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

PB9047
10209-274EA 808.99 USD InStock
10209-274
Anti-Cathepsin D Polyclonal Antibody
Antibodies
Polyclonal antibody for Cathepsin D/CTSD detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Cathepsin D/CTSD information: Molecular Weight: 44552 MW; Subcellular Localization: Lysosome. Melanosome. Secreted, extracellular space. Identified by mass spectrometry in melanosome fractions from stage I to stage IV. In aortic samples, detected as an extracellular protein loosely bound to the matrix (PubMed:20551380); Tissue Specificity: Expressed in the aorta extrcellular space (at protein level).

Type: Primary
Antigen: Cathepsin D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty