Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tff1 Polyclonal Antibody

Supplier: Boster
A01391-1
76171-424EA 622.93 USD InStock
76171-424
Anti-Tff1 Polyclonal Antibody
Antibodies
Polyclonal antibody for PS2/Tff1 detection. Host: Rabbit.Size: 100ug/vial. Tested applications: ELISA. Reactive species: Mouse. PS2/Tff1 information: Molecular Weight: 9670 MW; Subcellular Localization: Secreted; Tissue Specificity: Strong expression found in normal gastric mucosa. Not detected in the small or large intestine nor in normal mammary gland.

Type: Primary
Antigen: Tff1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty