Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-12 Lipoxygenase Polyclonal Antibody

Supplier: Boster
PA1485-1
76172-880EA 718.26 USD InStock
76172-880
Anti-12 Lipoxygenase Polyclonal Antibody
Antibodies
Polyclonal antibody for 12 LIPOXYGENASE/ALOX12 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. 12 LIPOXYGENASE/ALOX12 information: Molecular Weight: 25762 MW; Subcellular Localization: Ubiquitin: Cytoplasm . Nucleus ; Tissue Specificity: Expressed in vascular smooth muscle cells.

Type: Primary
Antigen: 12 Lipoxygenase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty