Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL-18 Polyclonal Antibody

Supplier: Boster
PB9245
76172-964EA 622.93 USD InStock
76172-964
Anti-IL-18 Polyclonal Antibody
Antibodies
Polyclonal antibody for IL 1F4/IL18 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Mouse. IL 1F4/IL18 information: Molecular Weight: 22135 MW; Subcellular Localization: Secreted.

Type: Primary
Antigen: IL-18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty