Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Caveolin-2 Polyclonal Antibody

Supplier: Boster
PB9166
76173-208EA 698.31 USD InStock
76173-208
Anti-Caveolin-2 Polyclonal Antibody
Antibodies
Polyclonal antibody for CAVEOLIN 2/CAV2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IF. Reactive species: Human. CAVEOLIN 2/CAV2 information: Molecular Weight: 18291 MW; Subcellular Localization: Nucleus. Cytoplasm. Golgi apparatus membrane; Peripheral membrane protein. Cell membrane; Peripheral membrane protein. Membrane, caveola; Peripheral membrane protein. Potential hairpin-like structure in the membrane. Membrane protein of caveolae. Tyr-19-phosphorylated form is enriched at sites of cell-cell contact and is translocated to the nucleus in complex with MAPK1 in response to insulin (By similarity). Tyr-27- phosphorylated form is located both in the cytoplasm and plasma membrane. CAV1-mediated Ser-23-phosphorylated form locates to the plasma membrane. Ser-36-phosphorylated form resides in intracellular compartments; Tissue Specificity: Expressed in endothelial cells, smooth muscle cells, skeletal myoblasts and fibroblasts.

Type: Primary
Antigen: Caveolin-2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty