Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EDA/Eda A2 Polyclonal Antibody

Supplier: Boster
PB9191
76173-248EA 622.93 USD InStock
76173-248
Anti-EDA/Eda A2 Polyclonal Antibody
Antibodies
Polyclonal antibody for EDA A2/EDA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. EDA A2/EDA information: Molecular Weight: 41294 MW; Subcellular Localization: Cell membrane; Single-pass type II membrane protein; Tissue Specificity: Not abundant; expressed in specific cell types of ectodermal (but not mesodermal) origin of keratinocytes, hair follicles, sweat glands. Also in adult heart, liver, muscle, pancreas, prostate, fetal liver, uterus, small intestine and umbilical chord.

Type: Primary
Antigen: EDA/Eda A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty