Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HOXA11 Polyclonal Antibody

Supplier: Boster
PB9218
76173-302EA 698.31 USD InStock
76173-302
Anti-HOXA11 Polyclonal Antibody
Antibodies
Polyclonal antibody for HOXA11 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. HOXA11 information: Molecular Weight: 34486 MW; Subcellular Localization: Nucleus.

Type: Primary
Antigen: HOXA11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty