Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TFRC Polyclonal Antibody

Supplier: Boster
PB9233
76173-332EA 698.31 USD InStock
76173-332
Anti-TFRC Polyclonal Antibody
Antibodies
Polyclonal antibody for TRANSFERRIN RECEPTOR/TFRC detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. TRANSFERRIN RECEPTOR/TFRC information: Molecular Weight: 84871 MW; Subcellular Localization: Cell membrane ; Single-pass type II membrane protein . Melanosome . Identified by mass spectrometry in melanosome fractions from stage I to stage IV.

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty