Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABCB4/Mdr3 Polyclonal Antibody

Supplier: Boster
PB9275
76173-384EA 698.31 USD InStock
76173-384
Anti-ABCB4/Mdr3 Polyclonal Antibody
Antibodies
Polyclonal antibody for MDR3/ABCB4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. MDR3/ABCB4 information: Molecular Weight: 141523 MW; Subcellular Localization: Cell membrane; Multi-pass membrane protein.

Type: Primary
Antigen: ABCB4/Mdr3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty