Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP-14 Polyclonal Antibody

Supplier: Boster

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

PB9283
76173-400EA 622.93 USD InStock
76173-400
Anti-MMP-14 Polyclonal Antibody
Antibodies
Polyclonal antibody for MT1 MMP/MMP14 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. MT1 MMP/MMP14 information: Molecular Weight: 65894 MW; Subcellular Localization: Membrane ; Single-pass type I membrane protein . Melanosome. Cytoplasm. Identified by mass spectrometry in melanosome fractions from stage I to stage IV. Forms a complex with BST2 and localizes to the cytoplasm; Tissue Specificity: Expressed in stromal cells of colon, breast, and head and neck. Expressed in lung tumors.

Type: Primary
Antigen: MMP-14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty