Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neuropilin-1 Polyclonal Antibody

Supplier: Boster
PB9300
76173-434EA 698.31 USD InStock
76173-434
Anti-Neuropilin-1 Polyclonal Antibody
Antibodies
Polyclonal antibody for NEUROPILIN 1/NRP1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. NEUROPILIN 1/NRP1 information: Molecular Weight: 103134 MW; Subcellular Localization: Cell membrane; Single-pass type I membrane protein; Tissue Specificity: The expression of isoforms 1 and 2 does not seem to overlap. Isoform 1 is expressed by the blood vessels of different tissues. In the developing embryo it is found predominantly in the nervous system. In adult tissues, it is highly expressed in heart and placenta; moderately in lung, liver, skeletal muscle, kidney and pancreas; and low in adult brain. Isoform 2 is found in liver hepatocytes, kidney distal and proximal tubules.

Type: Primary
Antigen: Neuropilin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty