Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P2X5 Polyclonal Antibody

Supplier: Boster
PB9305
76173-444EA 698.31 USD InStock
76173-444
Anti-P2X5 Polyclonal Antibody
Antibodies
Polyclonal antibody for P2X5/P2RX5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. P2X5/P2RX5 information: Molecular Weight: 47205 MW; Subcellular Localization: Membrane; Multi-pass membrane protein; Tissue Specificity: Expressed at high levels in brain and immune system.

Type: Primary
Antigen: P2X5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty