Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PAPP A Polyclonal Antibody

Supplier: Boster
PB9306
76173-446EA 698.31 USD InStock
76173-446
Anti-PAPP A Polyclonal Antibody
Antibodies
Polyclonal antibody for PAPPALYSIN 1/PAPPA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. PAPPALYSIN 1/PAPPA information: Molecular Weight: 180973 MW; Subcellular Localization: Secreted ; Tissue Specificity: High levels in placenta and pregnancy serum. In placenta, expressed in X cells in septa and anchoring villi, and in syncytiotrophoblasts in the chorionic villi. Lower levels are found in a variety of other tissues including kidney, myometrium, endometrium, ovaries, breast, prostate, bone marrow, colon, fibroblasts and osteoblasts.

Type: Primary
Antigen: PAPP A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty