Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PKC Alpha Polyclonal Antibody

Supplier: Boster
PB9318
76173-470EA 622.92 USD InStock
76173-470
Anti-PKC Alpha Polyclonal Antibody
Antibodies
Polyclonal antibody for PKC alpha/PRKCA detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. PKC alpha/PRKCA information: Molecular Weight: 76750 MW; Subcellular Localization: Cytoplasm. Cell membrane; Peripheral membrane protein. Mitochondrion membrane ; Peripheral membrane protein . Nucleus.

Type: Primary
Antigen: PKC Alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty