Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ALOX5/5 Lo Polyclonal Antibody

Supplier: Boster
RP1031
76174-866EA 837.99 USD InStock
76174-866
Anti-ALOX5/5 Lo Polyclonal Antibody
Antibodies
Polyclonal antibody for 5 LO/ALOX5 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. 5 LO/ALOX5 information: Molecular Weight: 77983 MW; Subcellular Localization: Cytoplasm. Nucleus matrix. Nucleus membrane; Peripheral membrane protein. Shuttles between cytoplasm and nucleus. Found exclusively in the nucleus, when phosphorylated on Ser-272. Calcium binding promotes translocation from the cytosol and the nuclear matrix to the nuclear envelope and membrane association.

Type: Primary
Antigen: ALOX5/5 Lo
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty