Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Collagen III Polyclonal Antibody

Supplier: Boster
RP1033
76174-870EA 718.26 USD InStock
76174-870
Anti-Collagen III Polyclonal Antibody
Antibodies
Polyclonal antibody for COLLAGEN III/COL3A1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. COLLAGEN III/COL3A1 information: Molecular Weight: 138564 MW; Subcellular Localization: Secreted, extracellular space, extracellular matrix .

Type: Primary
Antigen: Collagen III
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty