Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD59 Polyclonal Antibody

Supplier: Boster
RP1076
76174-942EA 837.99 USD InStock
76174-942
Anti-CD59 Polyclonal Antibody
Antibodies
Polyclonal antibody for CD59 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CD59 information: Molecular Weight: 14177 MW; Subcellular Localization: Cell membrane; Lipid-anchor, GPI-anchor. Secreted. Soluble form found in a number of tissues.

Type: Primary
Antigen: CD59
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty