Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JNK2 Polyclonal Antibody

Supplier: Boster
RP1077
76174-944EA 718.26 USD InStock
76174-944
Anti-JNK2 Polyclonal Antibody
Antibodies
Polyclonal antibody for JNK2/MAPK9 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. JNK2/MAPK9 information: Molecular Weight: 48139 MW; Subcellular Localization: Cytoplasm . Nucleus .

Type: Primary
Antigen: JNK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty