Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FABP4/A Fabp Polyclonal Antibody

Supplier: Boster
RP1085
76174-958EA 837.99 USD InStock
76174-958
Anti-FABP4/A Fabp Polyclonal Antibody
Antibodies
Polyclonal antibody for A FABP/FABP4 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. A FABP/FABP4 information: Molecular Weight: 14719 MW; Subcellular Localization: Cytoplasm. Nucleus. Depending on the nature of the ligand, a conformation change exposes a nuclear localization motif and the protein is transported into the nucleus. Subject to constitutive nuclear export (By similarity).

Type: Primary
Antigen: FABP4/A Fabp
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty