Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Horse Equine FGF9 PicoKine; ELISA Kit, Boster

Supplier: Boster
PicoKine
EK0348-EQ
76171-924EA 850.33 USD InStock
76171-924
Horse Equine FGF9 PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich High Sensitivity ELISA kit for Quantitative Detection of horse equine FGF9

  • Sensitivity, Detect single digit picogram level of target protein
  • Specificity, Thoroughly tested for cross-reactivity with family members
  • Stability, Guaranteed to remain stable for up to 12 months with proper storage
  • Selection, 1000+ kits available, covering most common ELISA targets
  • Service, On-demand technical support directly from ELISA experts

Detect Horse FGF9 with <15pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: serum, plasma(heparin, EDTA)and cell culture supernates. This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. FGF9 tissue specificity: Glial cells.

96wells/kit, with removable strips.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty