Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NAMPT IgG Polyclonal Antibody

Supplier: Boster
PB10009
76172-900EA 698.31 USD InStock
76172-900
Anti-NAMPT IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Nicotinamide phosphoribosyltransferase(NAMPT) detection. Tested with WB, IHC-P, ELISA in Human;Mouse;Rat.

1. 'Entrez Gene: PBEF1 pre-B-cell colony enhancing factor 1'.
2. Samal B, Sun Y, Stearns G, Xie C, Suggs S, McNiece I (February 1994). 'Cloning and characterization of the cDNA encoding a novel human pre-B-cell colony-enhancing factor'. Mol. Cell. Biol. 14 (2): 1431–7.
3. Wang T, Zhang X, Bheda P, Revollo JR, Imai S, Wolberger C (July 2006). 'Structure of Nampt/PBEF/visfatin, a mammalian NAD+ biosynthetic enzyme'. Nat. Struct. Mol. Biol. 13 (7): 661–2.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: NAMPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty