Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VAMP7 IgG Polyclonal Antibody

Supplier: Boster
PB10017
76172-916EA 622.76 USD InStock
76172-916
Anti-VAMP7 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Vesicle-associated membrane protein 7(VAMP7) detection. Tested with WB in Human;Rat.

1. 'Entrez Gene: SYBL1 synaptobrevin-like 1'.
2. D'Esposito M, Ciccodicola A, Gianfrancesco F, Esposito T, Flagiello L, Mazzarella R, Schlessinger D, D'Urso M (Jul 1996). "A synaptobrevin-like gene in the Xq28 pseudoautosomal region undergoes X inactivation". Nat. Genet. 13 (2): 227–9.
3. Filippini F, Rossi V, Galli T, Budillon A, D'Urso M, D'Esposito M (Jul 2001). 'Longins: a new evolutionary conserved VAMP family sharing a novel SNARE domain'. Trends Biochem. Sci. 26 (7): 407–9.

Add 0.2 ml of distilled water will yield a concentration of 500 ug/ml.

Type: Primary
Antigen: VAMP7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty