Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WNT1 Rabbit Polyclonal Antibody

Supplier: Boster
PB9460
76172-982EA 622.93 USD InStock
76172-982
Anti-WNT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Proto-oncogene Wnt-1(WNT1) detection. Tested with WB in Human.

Wingless-type MMIV integration site family, member 1 is a protein that in humans is encoded by the WNT1 gene. This gene is a member of the WNT gene family. The gene was assigned to human chromosome 12q13.12. The WNT gene family consists of structurally related genes that encode secreted signaling proteins. The Wnt1 protein functions in the induction of the mesencephalon and cerebellum. However, the gene mutations might not have a significant role in Joubert syndrome.

Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

WB: The detection limit for Wnt1 is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: WNT2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty