Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SERPINB5 Rabbit Polyclonal Antibody

Supplier: Boster
PB9356
76173-546EA 698.31 USD InStock
76173-546
Anti-SERPINB5 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Serpin B5(SERPINB5) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.

SERPINB5 is also known as PI5 or maspin. Maspin (mammary serine protease inhibitor) is a protein that in humans is encoded by the SERPINB5 gene. Maspin is expressed in the skin, prostate, testis, intestine, tongue, lung, and the thymus. Maspin is a member of the serpin superfamily of serine protease inhibitors.[1] The primary function of most members of this family is to regulate the breakdown of proteins by inhibiting the catalytic activity of proteinases. Through this mechanism of action, serpins regulate a number of cellular processes includingphagocytosis, coagulation, and fibrinolysis.

Add 0.2 ml of distilled water will yield a concentration of 500ug/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

WB: The detection limit for MASPIN is approximately 0.1 ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty