Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SFRP2 IgG Polyclonal Antibody

Supplier: Boster
PB9362
76173-558EA 622.93 USD InStock
76173-558
Anti-SFRP2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Secreted frizzled-related protein 2(SFRP2) detection. Tested with WB, ELISA in Human.

SFRP2 is also known as FRP-2, SARP1 or SDF-5. It is Expressed in adipose tissue, heart, brain, skeletal muscle, pancreas, thymus, prostate, testis, ovary, small intestine and colon. Highest levels in adipose tissue, small intestine and colon. This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer. 

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Each vial contains 5 mg BSA, 0.9mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.

WB: The detection limit for SFRP2 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SELP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Yes
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty