Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RUNX3 Rabbit Polyclonal Antibody

Supplier: Boster
PB9432
76173-696EA 698.31 USD InStock
76173-696
Anti-RUNX3 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Runt-related transcription factor 3(RUNX3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Runt-related transcription factor 3, also called A ml2 is a protein that in humans is encoded by the RUNX3 gene. The RUNX3 gene encodes a Runt-related transcription factor, which is part of the RUNX gene family. By fluorescence in situ hybridization, RUNX3 was assigned to human chromosome 1p36.11. RUNX3 binds to the core site of murine Leukemia virus, the core sequences in the enhancer of the polyomavirus, and also to the enhancers of the T-cell receptor genes. It may be involved in the control of cellular proliferation and/or differentiation.

Add 0.2 ml of distilled water will yield a concentration of 500µg/ ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na2HPO₄, 0.05 mg NaN₃.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: SDHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty