Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VDAC1 IgG Polyclonal Antibody

Supplier: Boster
PB9455
76173-740EA 698.31 USD InStock
76173-740
Anti-VDAC1 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Voltage-dependent anion-selective channel protein 1(VDAC1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

The voltage-dependent anion channel (VDAC) of the outer mitochondrial membrane is a small, abundant outer membrane pore-forming protein found in the outer membranes of all eukaryotic mitochondria. The VDAC protein is thought to form the major pathway for movement of adenine nucleotides through the outer membrane and to be the mitochondrial binding site for hexokinase and glycerol kinase. At low transmembrane voltage, VDAC is open for anions such as phosphate, chloride, and adenine nucleotides. At higher transmembrane voltage, VDAC functions as a selective channel for cations and uncharged molecules. These features make VDAC likely to play a role in mitochondrial energy metabolism. Huizing et al. studied by Northern and Western blot analyses the human tissue distribution of mitochondrial transmembrane metabolite carriers. They found that VDAC1 mRNA has a ubiquitous distribution, with most pronounced expression in heart, liver, and skeletal muscle, whereas the VDAC2 isoform appears to be expressed only in the heart.

Type: Primary
Antigen: VDAC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty